Anti-CLCN3 Mouse monoclonal antibody [clone: S258-5]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134285-100EA 3949.41 DKK
ABCAAB134285-100
Anti-CLCN3 Mouse monoclonal antibody [clone: S258-5]
Type: Primary
Antigen: CLCN3 (chloride channel, voltage-sensitive 3)
Clonality: monoclonal
Clone: S258-5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR