Anti-GRM5 Mouse monoclonal antibody [clone: S75-33]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB134271-100EA 0 DKK
ABCAAB134271-100
Anti-GRM5 Mouse monoclonal antibody [clone: S75-33]
Type: Primary
Antigen: GRM5 (glutamate receptor, metabotropic 5)
Clonality: monoclonal
Clone: S75-33
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat