Anti-Indoleamine 2, 3-dioxygenase Goat Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134197-100EA 4100.24 DKK
ABCAAB134197-100
Anti-Indoleamine 2, 3-dioxygenase Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Indoleamine 2, 3-dioxygenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR