Anti-HSD17B1 Rabbit Monoclonal Antibody [clone: EP1681Y]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134193-100EA 6080 DKK
ABCAAB134193-100
Anti-HSD17B1 Rabbit Monoclonal Antibody [clone: EP1681Y]
Antistoffer
Type: Primary
Antigen: HSD17B1 (hydroxysteroid (17-beta) dehydrogenase 1)
Clonality: monoclonal
Clone: EP1681Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR