Anti-ALDH1A1 Rabbit Monoclonal Antibody [clone: EP1932Y]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134188-100EA 3876.59 DKK
ABCAAB134188-100
Anti-ALDH1A1 Rabbit Monoclonal Antibody [clone: EP1932Y]
Antistoffer
Type: Primary
Antigen: ALDH1A1 (aldehyde dehydrogenase 1 family, member A1)
Clonality: monoclonal
Clone: EP1932Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR