Anti-MMP1 Rabbit Monoclonal Antibody [clone: EP1249Y]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB134184-100EA 3503.84 DKK
ABCAAB134184-100
Anti-MMP1 Rabbit Monoclonal Antibody [clone: EP1249Y]
Antistoffer
Type: Primary
Antigen: MMP1 (matrix metallopeptidase 1 (interstitial collagenase))
Clonality: monoclonal
Clone: EP1249Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu