Anti-BIRC5 Rabbit Monoclonal Antibody [clone: EPR2675]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134170-100EA 6790 DKK
ABCAAB134170-100
Anti-BIRC5 Rabbit Monoclonal Antibody [clone: EPR2675]
Antistoffer
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: monoclonal
Clone: EPR2675
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR