Anti-TPBG Rabbit Monoclonal Antibody [clone: EPR5529]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134162-100EA 6590 DKK
ABCAAB134162-100
Anti-TPBG Rabbit Monoclonal Antibody [clone: EPR5529]
Antistoffer
Type: Primary
Antigen: TPBG (trophoblast glycoprotein)
Clonality: monoclonal
Clone: EPR5529
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR