Anti-AKT1S1 Rabbit Monoclonal Antibody [clone: EPR6262]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134158-100EA 3615.67 DKK
ABCAAB134158-100
Anti-AKT1S1 Rabbit Monoclonal Antibody [clone: EPR6262]
Antistoffer
Type: Primary
Antigen: AKT1S1 (AKT1 substrate 1 (proline-rich))
Clonality: monoclonal
Clone: EPR6262
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR