Anti-CD41 Rabbit Monoclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB134131-100EA 6930 DKK
ABCAAB134131-100
Anti-CD41 Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu