Anti-SEMA4D Rabbit Monoclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB134128-100EA 6690 DKK
ABCAAB134128-100
Anti-SEMA4D Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu