Anti-CD79B Rabbit Monoclonal Antibody [clone: EPR6860]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134103-100EA 4323.89 DKK
ABCAAB134103-100
Anti-CD79B Rabbit Monoclonal Antibody [clone: EPR6860]
Antistoffer
Type: Primary
Antigen: CD79B (CD79b molecule, immunoglobulin-associated beta)
Clonality: monoclonal
Clone: EPR6860
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR