Anti-ADO Rabbit Monoclonal Antibody [clone: EPR6581]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134102-100EA 3615.67 DKK
ABCAAB134102-100
Anti-ADO Rabbit Monoclonal Antibody [clone: EPR6581]
Antistoffer
Type: Primary
Antigen: ADO (2-aminoethanethiol (cysteamine) dioxygenase)
Clonality: monoclonal
Clone: EPR6581
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR