Anti-IRS2 Rabbit Monoclonal Antibody [clone: EPR904]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB134101-100EA 6630 DKK
ABCAAB134101-100
Anti-IRS2 Rabbit Monoclonal Antibody [clone: EPR904]
Antistoffer
Type: Primary
Antigen: IRS2 (insulin receptor substrate 2)
Clonality: monoclonal
Clone: EPR904
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu