Anti-RET Rabbit Monoclonal Antibody [clone: EPR2871]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134100-100EA 4137.51 DKK
ABCAAB134100-100
Anti-RET Rabbit Monoclonal Antibody [clone: EPR2871]
Antistoffer
Type: Primary
Antigen: RET (ret proto-oncogene)
Clonality: monoclonal
Clone: EPR2871
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR