Anti-GAP43 Rabbit monoclonal antibody unconjugated [clone: EPR5031]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134075-100EA 4062.96 DKK
ABCAAB134075-100
Anti-GAP43 Rabbit monoclonal antibody unconjugated [clone: EPR5031]
Antistoffer
Type: Primary
Antigen: GAP43 (growth associated protein 43)
Clonality: monoclonal
Clone: EPR5031
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR