Anti-STUB1 Rabbit Monoclonal Antibody [clone: EPR4447]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB134064-100EA 7200 DKK
ABCAAB134064-100
Anti-STUB1 Rabbit Monoclonal Antibody [clone: EPR4447]
Antistoffer
Type: Primary
Antigen: STUB1 (STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: EPR4447
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR