Anti-USH1C Rabbit Monoclonal Antibody [clone: EPR8131]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133763-100EA 3688.6 DKK
ABCAAB133763-100
Anti-USH1C Rabbit Monoclonal Antibody [clone: EPR8131]
Antistoffer
Type: Primary
Antigen: USH1C (Usher syndrome 1C (autosomal recessive, severe))
Clonality: monoclonal
Clone: EPR8131
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR