Anti-CUTC Rabbit Monoclonal Antibody [clone: EPR7929]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133762-100EA 5910 DKK
ABCAAB133762-100
Anti-CUTC Rabbit Monoclonal Antibody [clone: EPR7929]
Antistoffer
Type: Primary
Antigen: CUTC (cutC copper transporter)
Clonality: monoclonal
Clone: EPR7929
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR