Anti-STX3 Rabbit Monoclonal Antibody [clone: EPR8543]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133750-100EA 7090 DKK
ABCAAB133750-100
Anti-STX3 Rabbit Monoclonal Antibody [clone: EPR8543]
Antistoffer
Type: Primary
Antigen: STX3 (syntaxin 3)
Clonality: monoclonal
Clone: EPR8543
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR