Anti-DPC Rabbit Monoclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB133652-100EA 6630 DKK
ABCAAB133652-100
Anti-DPC Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: DPC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu