Anti-RENT1/HUPF1 Rabbit Monoclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133564-100EA 6590 DKK
ABCAAB133564-100
Anti-RENT1/HUPF1 Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: RENT1/HUPF1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR