Anti-CHK2 Rabbit Monoclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB133505-100EA 7150 DKK
ABCAAB133505-100
Anti-CHK2 Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: CHK2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu