Anti-IQGAP1 Rabbit Monoclonal Antibody
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB133490-100EA 7350 DKK
ABCAAB133490-100
Anti-IQGAP1 Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: IQGAP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestil nu