Anti-COX2 / Cyclooxygenase 2 Rabbit Monoclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133466-100EA 0 DKK
ABCAAB133466-100
Anti-COX2 / Cyclooxygenase 2 Rabbit Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: COX2 / Cyclooxygenase 2
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR