Anti-MUC16 Rabbit polyclonal antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133419-50EA 5100 DKK
ABCAAB133419-50
Anti-MUC16 Rabbit polyclonal antibody
Type: Primary
Antigen: MUC16 (mucin 16, cell surface associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR