Anti-RPA1 Rabbit Monoclonal Antibody [clone: EPR3473]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133364-100EA 6440 DKK
ABCAAB133364-100
Anti-RPA1 Rabbit Monoclonal Antibody [clone: EPR3473]
Antistoffer
Type: Primary
Antigen: RPA1 (replication protein A1, 70kDa)
Clonality: monoclonal
Clone: EPR3473
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR