Anti-TCF7L1 Rabbit Monoclonal Antibody [clone: EPR2031]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133360-100EA 5740 DKK
ABCAAB133360-100
Anti-TCF7L1 Rabbit Monoclonal Antibody [clone: EPR2031]
Antistoffer
Type: Primary
Antigen: TCF7L1 (transcription factor 7-like 1 (T-cell specific, HMG-box))
Clonality: monoclonal
Clone: EPR2031
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR