Anti-SUMO1 Rabbit Monoclonal Antibody [clone: EP298]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB133352-100EA 7250 DKK
ABCAAB133352-100
Anti-SUMO1 Rabbit Monoclonal Antibody [clone: EP298]
Antistoffer
Type: Primary
Antigen: SUMO1 (small ubiquitin-like modifier 1)
Clonality: monoclonal
Clone: EP298
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu