Anti-PRDX6 Rabbit Monoclonal Antibody [clone: EPR3754]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133348-100EA 6640 DKK
ABCAAB133348-100
Anti-PRDX6 Rabbit Monoclonal Antibody [clone: EPR3754]
Antistoffer
Type: Primary
Antigen: PRDX6 (peroxiredoxin 6)
Clonality: monoclonal
Clone: EPR3754
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR