Anti-SOX2 Rabbit Monoclonal Antibody [clone: EPR3130]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133337-100EA 4100.24 DKK
ABCAAB133337-100
Anti-SOX2 Rabbit Monoclonal Antibody [clone: EPR3130]
Antistoffer
Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: monoclonal
Clone: EPR3130
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR