Anti-PDCD6 Rabbit Monoclonal Antibody [clone: EPR3260]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133326-100EA 4100.24 DKK
ABCAAB133326-100
Anti-PDCD6 Rabbit Monoclonal Antibody [clone: EPR3260]
Antistoffer
Type: Primary
Antigen: PDCD6 (programmed cell death 6)
Clonality: monoclonal
Clone: EPR3260
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR