Anti-MCM2 Rabbit Monoclonal Antibody [clone: EPR3727]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133325-100EA 3576.83 DKK
ABCAAB133325-100
Anti-MCM2 Rabbit Monoclonal Antibody [clone: EPR3727]
Antistoffer
Type: Primary
Antigen: MCM2 (minichromosome maintenance complex component 2)
Clonality: monoclonal
Clone: EPR3727
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR