Anti-COX1 / Cyclooxygenase 1 Rabbit Monoclonal Antibody [clone: EPR5867]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133319-100EA 6690 DKK
ABCAAB133319-100
Anti-COX1 / Cyclooxygenase 1 Rabbit Monoclonal Antibody [clone: EPR5867]
Antistoffer
Type: Primary
Antigen: COX1 / Cyclooxygenase 1
Clonality: monoclonal
Clone: EPR5867
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR