Anti-CALB2 Rabbit monoclonal antibody unconjugated [clone: EPR1799]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133316-100EA 4100.24 DKK
ABCAAB133316-100
Anti-CALB2 Rabbit monoclonal antibody unconjugated [clone: EPR1799]
Antistoffer
Type: Primary
Antigen: CALB2 (calbindin 2)
Clonality: monoclonal
Clone: EPR1799
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR