Anti-FEN1 Rabbit Monoclonal Antibody [clone: EPR4459]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB133311-100EA 6690 DKK
ABCAAB133311-100
Anti-FEN1 Rabbit Monoclonal Antibody [clone: EPR4459]
Antistoffer
Type: Primary
Antigen: FEN1 (flap structure-specific endonuclease 1)
Clonality: monoclonal
Clone: EPR4459
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu