Anti-A2M Rabbit Monoclonal Antibody [clone: EPR4431]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133299-100EA 6780 DKK
ABCAAB133299-100
Anti-A2M Rabbit Monoclonal Antibody [clone: EPR4431]
Antistoffer
Type: Primary
Antigen: A2M (alpha-2-macroglobulin)
Clonality: monoclonal
Clone: EPR4431
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR