Anti-APOF Rabbit monoclonal antibody unconjugated [clone: EP2909]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133295-100EA 6020 DKK
ABCAAB133295-100
Anti-APOF Rabbit monoclonal antibody unconjugated [clone: EP2909]
Antistoffer
Type: Primary
Antigen: APOF (apolipoprotein F)
Clonality: monoclonal
Clone: EP2909
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR