Anti-AVEN Rabbit Monoclonal Antibody [clone: EPR4720]
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB133285-100EA 5990 DKK
ABCAAB133285-100
Anti-AVEN Rabbit Monoclonal Antibody [clone: EPR4720]
Antistoffer
Type: Primary
Antigen: AVEN (apoptosis, caspase activation inhibitor)
Clonality: monoclonal
Clone: EPR4720
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu