Anti-CCNE1 Rabbit monoclonal antibody unconjugated [clone: EPR194]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133266-100EA 3988.41 DKK
ABCAAB133266-100
Anti-CCNE1 Rabbit monoclonal antibody unconjugated [clone: EPR194]
Antistoffer
Type: Primary
Antigen: CCNE1 (cyclin E1)
Clonality: monoclonal
Clone: EPR194
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR