Anti-DFFA Rabbit monoclonal antibody unconjugated [clone: EPR4221]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133250-100EA 3763.12 DKK
ABCAAB133250-100
Anti-DFFA Rabbit monoclonal antibody unconjugated [clone: EPR4221]
Antistoffer
Type: Primary
Antigen: DFFA (DNA fragmentation factor, 45kDa, alpha polypeptide)
Clonality: monoclonal
Clone: EPR4221
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR