Anti-LRRFIP1 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB133218-50EA 4100.24 DKK
ABCAAB133218-50
Anti-LRRFIP1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: LRRFIP1 (leucine rich repeat (in FLII) interacting protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR