Anti-HDAC4/5/9 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB131524-100EA 6150 DKK
ABCAAB131524-100
Anti-HDAC4/5/9 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: HDAC4/5/9 (histone deacetylase 4/5/9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat