Anti-CCNB1 Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ABCAAB131502-100EA 6630 DKK
ABCAAB131502-100
Anti-CCNB1 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CCNB1 (cyclin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human