Anti-PTPN6 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131500-100EA 4100.24 DKK
ABCAAB131500-100
Anti-PTPN6 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PTPN6 (protein tyrosine phosphatase, non-receptor type 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR