Anti-CD38 Mouse Monoclonal Antibody [clone: B307 (AT1)]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131420-200EA 6450 DKK
ABCAAB131420-200
Anti-CD38 Mouse Monoclonal Antibody [clone: B307 (AT1)]
Antistoffer
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: monoclonal
Clone: B307 (AT1)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR