Anti-ANK2 Mouse monoclonal antibody [clone: S105-13]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131419-100EA 5280 DKK
ABCAAB131419-100
Anti-ANK2 Mouse monoclonal antibody [clone: S105-13]
Type: Primary
Antigen: ANK2 (ankyrin 2, neuronal)
Clonality: monoclonal
Clone: S105-13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR