Anti-MAPT Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131354-100EA 6630 DKK
ABCAAB131354-100
Anti-MAPT Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR