Anti-C1QBP Rabbit Monoclonal Antibody [clone: EPR8871]

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131284-100EA 3802.04 DKK
ABCAAB131284-100
Anti-C1QBP Rabbit Monoclonal Antibody [clone: EPR8871]
Antistoffer
Type: Primary
Antigen: C1QBP (complement component 1, q subcomponent binding protein)
Clonality: monoclonal
Clone: EPR8871
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR