Anti-MAP2K3 Rabbit Polyclonal Antibody

Leverandør: Abcam
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ABCAAB131283-100EA 4174.79 DKK
ABCAAB131283-100
Anti-MAP2K3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MAP2K3 (mitogen-activated protein kinase kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR